4-Methyl-2-(3-pyridinyl)-5-thiazolecarboxylic acid ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: 4-Methyl-2-(pyridin-3-yl)thiazole-5-carboxylic acid

H51723.03 H51723.06
H51723.03EA 82.8 GBP
H51723.03 H51723.06
4-Methyl-2-(3-pyridinyl)-5-thiazolecarboxylic acid ≥97%
4-Methyl-2-(3-pyridinyl)-5-thiazolecarboxylic acid

Purity: 97% CAS No.: 39091-01-5 Molecular Formula: C10H8N2O2S Formula Weight: 220.25 Melting Point: 244-248° MDL No.: MFCD00171753

Formula: C₁₀H₈N₂O₂S
MW: 220,25 g/mol
Melting Pt: 244…248 °C
Storage Temperature: Ambient
MDL Number: MFCD00171753
CAS Number: 39091-01-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR