CPC® Aseptiquik® W Series Genderless Connector to 1-1/2" Hose Barb, High Temperature, Polycarbonate, AQW17024HT; 1/PK
Supplier: Avantor Fluid Handling
VWR Item#MFLX20202-74
Your Organization's Price:
Retrieving
Provide easy, secure sterile connections even in nonsterile environments now in newly engineered larger sizes.
  • Genderless design simplifies system integration and eases single-use systems specifications with one part number for both halves
  • Intuitive four-step connection process reduces risk of operator error 'FLIP - CLICK - PULL - SNAP'
  • Secure side latches increase connector robustness for wide variety of applications
  • Membrane pull tabs ensure simultaneous and secure removal of both membranes
  • No secondary component needed; no need to purchase and store extra materials or tools to complete the sterile connection process
  • Move liquid media faster to increase process efficiency using largest AseptiQuik® flow diameter
All fittings can be sterilised by gamma irradiation up to 50 kGy. Use high-temperature fittings to sterilize via autoclave up to 266 °F (130 °C) for up to 60 minutes.