CPC® Aseptiquik® W Series Genderless Connector to 1" Hose Barb, High Temperature, Polycarbonate, AQW17016HT; 1/PK
Supplier: Avantor Fluid Handling
Provide easy, secure sterile connections even in nonsterile environments now in newly engineered larger sizes.
- Genderless design simplifies system integration and eases single-use systems specifications with one part number for both halves
- Intuitive four-step connection process reduces risk of operator error 'FLIP - CLICK - PULL - SNAP'
- Secure side latches increase connector robustness for wide variety of applications
- Membrane pull tabs ensure simultaneous and secure removal of both membranes
- No secondary component needed; no need to purchase and store extra materials or tools to complete the sterile connection process
- Move liquid media faster to increase process efficiency using largest AseptiQuik® flow diameter
All fittings can be sterilised by gamma irradiation up to 50 kGy. Use high-temperature fittings to sterilize via autoclave up to 266 °F (130 °C) for up to 60 minutes.