CPC® APC Quick-Disconnect Fitting, Elbow Hose Barb Insert, Acetal, Valved, 3/8" ID, APCD23006; 25/PK
Supplier: Avantor Fluid Handling
VWR Item#MFLX06353-34BLK
Your Organization's Price:
Retrieving
Fittings connect in a snap.
  • No exposed metal parts
  • Protects against accidental disconnects
These CPC® APC-Series quick-disconnect fittings, combine a compact design with durable acetal construction, making them an excellent choice for many applications. All fittings feature a 1/4” flow, plastic thumb latch, fewer moving parts, and a smooth contoured design to deliver ease-of-use and excellent flow in a compact size. Simply press the thumb latch to disconnect fitting halves. Choose from in-line compression and hose barb bodies which feature shrouded latches to provide further protection against accidental disconnect. The ferruleless PTF compression fittings do not require ferrules to achieve a secure connection, which makes them easier to use and reuse. PTF fittings are designed for semi-rigid tubing, i.e., polyethylene, nylon, polyurethane, etc.
A complete coupler requires a fitting body and an insert (order separately).
Minimun 25 pcs required to purchase