Fitting, White Polypropylene, Straight, Hose Barb Union, 4-5.25 mm ID; 100/Pk
Supplier: Avantor Fluid Handling
VWR Item#MFLX30601-00BLK
Your Organization's Price:
Retrieving
Quick and easy assembly without tools.
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Chemical resistant
  • Connection Type: 4-5.25 mm int.Ø