Smiths Medical® Fitting, Polycarbonate, Four-Way Stopcock, Male Slip Luer, Non-Sterile; 1000/Pk
Supplier: Avantor Fluid Handling
VWR Item#MFLX30600-03BLK
Your Organization's Price:
Retrieving
Regulate flow without any leaking.
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Four-way stopcock/flow diverter with male luer slip connections
  • HDPE handle rotates 360°
Choose stopcocks with either a male luer lock or a male luer slip. Handle rotates 90° on one-way stopcocks, 180° on three-way stopcocks, and 360° on four-way stopcocks. All have PC body and HDPE stem.
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.