Masterflex® Ismatec® Microbore Tubing Fitting, size 14 (0.38mm ID) tubing, 316 SS, 2/pk
Supplier: Avantor Fluid Handling
Connect even the smallest tubing with a leak-free seal from vacuum to pressure.
- Multiple styles to fit your needs
- Stainless steel fitting sized to match the Ismatec® sizes - from 09 to 26
<b>Delivery information: </b>Each kit includes 2 fittings.