Masterflex® L/S® Miniflex® OEM Pump Head, 3-Rollers, Dual-Channel
Supplier: Avantor Fluid Handling
VWR Item#MFLX77220-10
Your Organization's Price:
Retrieving
Pump head accepts continuous lengths of tubing for clean flow paths.
  • Flow range of 1.8 to 220 ml/min—flow rate depends on drove rpm, pump head, and tubing size
  • Pump head accepts continuous lengths of tubing for clean flow paths
  • Dual-channel model delivers synchronous flow from two separate channels; single-channel model delivers highest flow rate
  • Automatic tubing retention makes tube loading quick and easy
The Masterflex® Miniflex® pump heads feature a wide occlusion angle that prevents fluid backflow up to the rated pressure of the tubing. Self-lubricating thermoplastic polyester rollers are low friction and long wearing for smooth operation and long life. A compact overall size envelope makes these heads ideal for OEM applications and integration into fluid systems and machinery.
<B>Note:</B> Dual-channel model must have same size tubing loaded in both channels to ensure tubing retention.