Masterflex® Transfer Tubing, Reinforced Peroxide-Cured Silicone, 3/4" ID x 1-1/8" OD; 25 Ft
Supplier: Avantor Fluid Handling
Low compression formulation keeps its shape and cleanliness.
- Ideal for general-purpose, single-use, and extended fluid path applications
- Single-use custom lengths and configurations available to minimise downtime
- Tubing size: 3/4" ID x 1 1/8" OD
<B>Applications:</B> Demanding high-pressure pharmaceutical, biotechnology, aerospace, and food and beverage applications. <br /><B>Characteristics:</B> Excellent heat resistance, low compression set, odorless, nontoxic, low tackiness, translucent.<br /><B>Certifications:</B> Elastomer is FDA compliant and meets USP Class VI. Tubing is manufactured according to GMP. <br /><B>Temperature range:</B> -60 to 400 °F (-51 to 204 °C) <br /><B>Sterilize:</B> Autoclave or gamma irradiation.