Masterflex® Open Impeller Centrifugal Pump, 155 GPM/33 ft, 304 SS; 230/460V
Supplier: Avantor Fluid Handling
Suitable for suspended solids.
- Close-coupled design offers smallest possible footprint to save space
- Back-pullout construction allows for servicing of pump without disturbing inlet and outlet piping
- Top centerline discharge ensures self-venting of entrapped gases
- Maximum flow rate 155 GPM
- Maximum feet of head 33 ft
- Operates on 230/460 VAC
Typical applications include food process, cooling systems, beverage processing and pharmaceutical services. The open impeller design means the pump can easily handle suspended solids up to a 3/4" spherical shape. Pumps are non-overloading and rated for continuous duty throughout entire operating range for reliable performance and long life.
<b>Delivery information: </b>Includes conduit box for hard-wiring.