Masterflex® FDA-Compliant Sanitary Drum Pump, 15 GPM, 39"L Inlet Tube, TEFC Motor; 220V
Supplier: Avantor Fluid Handling
VWR Item#MFLX75219-55
Your Organization's Price:
Retrieving
Capable of pumping fluids with viscosities up to 1000 cps.
  • 316 SS construction with PTFE elastomers
  • Easy to clean with Tri-Clamp® fittings
  • Flow rate of 15 GPM, Powered by 220V TEFC motor
The sanitary drum pumps are engineered to transfer low-viscosity food products, pharmaceutical materials, and personal care solutions from drums, totes, and kettles. Wetted surfaces are constructed with FDA-compliant materials which provide an ideal solution for sanitary applications.
<b>Delivery information: </b>Includes drive and pump tube.