CPC® ChemQuik® Quick-Disconnect Coupling, In-Line Body, Valved, Polypropylene, 1/2" OD, CQHD06100108; 1/PK
Supplier: Avantor Fluid Handling
VWR Item#MFLX31603-01
Your Organization's Price:
Retrieving
All plastic construction for high-purity use.
  • 100% metal-free couplings are ideal for ultrapure fluids, deionized water, and aggressive chemical applications
These ChemQuik® quick-disconnect couplings provide easy single-handed disconnection. These couplings feature valves to provide flow shut-off. The all plastic construction and lubricant-free design enables use in the most demanding applications.
For a complete coupling, order one body and one insert of the same size.