Masterflex® FDA-Compliant Sanitary Drum Pump, 26 GPM, 39"L Inlet Tube, Air Motor
Supplier: Avantor Fluid Handling
Capable of pumping fluids with viscosities up to 1000 cps.
- 316 SS construction with PTFE elastomers
- Easy to clean with Tri-Clamp® fittings
- Flow rate of 26 Powered by air motor
The sanitary drum pumps are engineered to transfer low-viscosity food products, pharmaceutical materials, and personal care solutions from drums, totes, and kettles. Wetted surfaces are constructed with FDA-compliant materials which provide an ideal solution for sanitary applications.
<b>Delivery information: </b>Includes drive and pump tube.