Seal Kit for Piston Pump 74931-20; 1/Each
Supplier: Avantor Fluid Handling
VWR Item#MFLX74931-95
Your Organization's Price:
Retrieving
Designed for optimal operation in your Ismatec pumps.
  • Choose from a wide range of replacement parts and accessories
The overall length of Solva 95605-series stop tubing is 16" (406 mm). The legacy Ismatec stop tubing was 400 mm long.<br /><br />Order extension tubing under "Accessories". For technical assistance selecting tubing formulations and sizes, contact our Application Specialists.