High-Head Enclosed Motor PP Centrifugal Pump; 4.6 GPM/36.1 ft, 230V
Supplier: Avantor Fluid Handling
Twin bearing system handles thrust loads for extended service life.
- Quiet, energy efficient, and compact
- Internal cooling circuit reduces bearing temperature
- Enclosed motor for use in harsh environments
- All wetted parts are nonmetallic for excellent chemical resistance
High-head polypropylene pumps have a closed impeller design to significantly increase the output pressure. Polypropylene construction provides excellent chemical resistance in applications, such as filtration, where a high-head is needed.
<b>Delivery information: </b>Includes a 6-ft (1.8 m) cord with stripped ends.