High-Precision High-Pressure Piston Pump Seal Kit, Organic Seal Kit
Supplier: Avantor Fluid Handling
Designed for optimal operation in your Ismatec pumps.
- Choose from a wide range of replacement parts and accessories
The overall length of Solva 95605-series stop tubing is 16" (406 mm). The legacy Ismatec stop tubing was 400 mm long.<br /><br />Order extension tubing under "Accessories". For technical assistance selecting tubing formulations and sizes, contact our Application Specialists.