PTFE Manifold; 4 Outlets; 1/2" Inlet NPT(F)
Supplier: Avantor Fluid Handling
VWR Item#MFLX31521-11
Your Organization's Price:
Retrieving
Chemically-inert PTFE handles some of your most aggressive chemical solutions.
  • Constructed of durable PTFE