Gems Hinged Liquid Level Float Switch, Nylon; 1/2" NPT External Mounting
Supplier: Avantor Fluid Handling
Get longer life and more durability with hermetically sealed switches.
- Get longer life and more durability with hermetically sealed switches
These hinged float switches operate by a hinged magnet that swings closed and trips a reed switch. Easily install at any height in tank for a wide range of applications.
These PVDF (Kynar®) magnetic float switches swing closed and trip an internal reed switch. The PVDF material is ideal for high purity liquid applications and many acids. Install at any height in tanks for high or low level alarms or process control.