Masterflex® Transfer Tubing, PVC Reinforced, 2" ID x 2-1/2" OD; 100 Ft
Supplier: Avantor Fluid Handling
More durable than standard PVC.
- Ideal for general-purpose, single-use, and extended fluid path applications
- Single-use custom lengths and configurations available to minimise downtime
- Tubing size: 2" ID x 2 1/2" OD
Applications: High-pressure use, food processing, and beverage handling.<br />Characteristics: Good acid/alkali resistance and aging characteristics. White polyester yarn with orange tracer yarn embedded in a transparent PVC wall.<br />Certification: Resin is FDA-compliant (21 CFR 177.199) and meets USDA requirements. NSF-listed (standard 51) with 3A Sanitary Standards for dairy.<br />Temperature range: -5 to 180 °F (-21 to 82 °C). NSF temperature to 140 °F.<br />Sterilize: Ethylene oxide.