Masterflex® L/S® 2-Stop Precision Pump Tubing, PharMed® BPT, L/S 13; 8/Pk
Supplier: Avantor Fluid Handling
Protect your light-sensitive samples.
- Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
- Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
- Single-use custom lengths and configurations available to minimise downtime
- Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
- Tubing Size: L/S 13
<B>Applications</B>: Cell and tissue culture work, pharmaceutical, light-sensitive samples<br /><B>Characteristics</B>: Nontoxic, nonhemolytic, extremely low permeability, resists acids, bases, and oxidizing agents; heat-sealable, bondable, formable, opaque beige<br />For use with Masterflex® multichannel pump heads.