Masterflex® Transfer Tubing, Tygon® 2375, 3/8" ID x 1/2" OD; 50 Ft
Supplier: Avantor Fluid Handling
Won't allow particulate build-up.
- Ideal for general-purpose, single-use, and extended fluid path applications
- Single-use custom lengths and configurations available to minimise downtime
- Tubing size: 3/8" ID x 1/2" OD
These transparent tubes are ideal for use with highly aggressive chemicals – acids, bases, ketones, salts, and alcohols. Characteristics include plasticizer free, smoother inner surface, hydrophobic, minimal outgassing, safe disposal via incineration, and nontoxic. NSF 51 Standard compliant and manufactured according to GMP. Temperature range is -108 to 130 °F (-78 to 54 °C). Can be sterilized with ethylene oxide or gamma irradiation.