Masterflex® L/S® Analog Modular Drive Replacement Motor, 600 rpm, IP21; 230 VAC
Supplier: Avantor Fluid Handling
Make the most of your Masterflex drives replacement parts and other accessories.
- Choose replacement motors based on your Masterflex Modular System
Extend the life of your Masterflex pump with these replacement parts.
Compatible with the following Masterflex system: 07553-77. Controller catalog number is 07553-78.