FMI Series RH Valveless Piston Pump Head, Ceramic, 1/4", 1/4-28 (F) ports
Supplier: Avantor Fluid Handling
VWR Item#MFLX07117-69
Your Organization's Price:
Retrieving
Small displacement provides excellent precision.
  • Fully adjustable from zero to maximum
Valveless piston pumps are low maintenance and suffer fewer piston siezures or clogs. With a single moving part, there is less drift over time, offering millions of cycles of reliable, repeatable pumping.