Masterflex® I/P® Standard Pump Head, Replacement End Bell Assembly, PPS Housing, SS Rotor, I/P® 82
Supplier: Avantor Fluid Handling
Get the most our of your Masterflex® Pump Head with these replacement parts.
- Choose replacement parts suitable for your L/S® and I/P® pump head
- Suitable for use with I/P® 82 Standard pump head
- Constructed with durable Polyphenylene Sulfide (PPS) housing and stainless steel (SS) rotor
Replacement parts to keep your pump head operating even longer. Two end bell assemblies are required for a complete head assembly.