Thermoplastic Ethyl Vinyl Acetate Microbore Tubing, 0.030" ID x 0.090" OD; 100 ft
Supplier: Avantor Fluid Handling
Biologically stable, nontoxic, and non-contaminating.
Applications: Ideal for biological testing.<br /> Characteristics: Cross-linked tubing is biologically stable, nontoxic, and non-contaminating. Flexible Micro-Line™ tubing has virtually 100% elastic memory—returns to its original shape with gentle heating. Tensile strength is 2700 psi; durometer hardness is Shore-A 90. Translucent.<br /> Certification: Resin is USP 23 Class VI, and National Formulary Vol. 14 specifications.<br /> Temperature range: –29 to 180 °F (–34 to 82 °C)<br /> Sterilize: With autoclave, ethylene oxide, gamma irradiation, or dry heat