Masterflex® I/P® High-Performance Precision Pump Tubing, Gore™ STA-PURE™ PCS-Series, I/P 89; 24"/Pk
Supplier: Avantor Fluid Handling
VWR Item#MFLX96241-89
Your Organization's Price:
Retrieving
Long pump life and chemically compatible to a wide range of chemicals.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy, and reproducibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Lot-to-lot consistency provides superior accuracy and repeatability
  • I/P® 89 tubing has a 5/8” (15.88 mm) inner diameter
  • Accepts 5/8” hose barb for tubing connection
Masterflex® Gore™ STA-PURE™ PCS-Series pump tubing offers significantly longer life combined with virtually no spallation; high pressure ratings; and consistent, repeatable flow rates; even after hundreds of hours of use.<br /><br />This tubing is your ideal choice for high purity or long-running, critical pharmaceutical and biopharmaceutical processing applications. The patented composite platinum-cured silicone and ePTFE (polytetrafluoroethylene), results in a tube that offers high-flex durability for greater resistance to rupture—even at applications up to 100 psig. Long-term pump testing proves that Gore Sta-Pure Pump Tubing - Series PCS lasts over 1000 hours at 200 rpm, or over 6000 hours at 100 rpm. Its long life eliminates the need to rotate tubing through the pump head, saving time and reducing maintenance.<br /><br />I/P® Precision pump tubing is compatible with Masterflex® Standard and Easy-Load pump heads. I/P® High-Performance Precision pump tubing is compatible with Masterflex® High-Performance pump heads. <br /><br /><B>NOTE: </B> There is an engineering questionnaire that needs to be completed before the order can be processed.