Masterflex® Transfer Tubing, Tygon® 2375, 5/32" ID x 7/32" OD; 50 Ft
Supplier: Avantor Fluid Handling
VWR Item#MFLX95630-14
Your Organization's Price:
Retrieving
Won't allow particulate build-up.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Single-use custom lengths and configurations available to minimise downtime
  • Tubing size: 5/32" ID x 7/32" OD
These transparent tubes are ideal for use with highly aggressive chemicals – acids, bases, ketones, salts, and alcohols. Characteristics include plasticizer free, smoother inner surface, hydrophobic, minimal outgassing, safe disposal via incineration, and nontoxic. NSF 51 Standard compliant and manufactured according to GMP. Temperature range is -108 to 130 °F (-78 to 54 °C). Can be sterilized with ethylene oxide or gamma irradiation.