Masterflex® L/S® Precision Pump Tubing, Versilon™ 2001 Tubing, L/S 18; 50 ft
Supplier: Avantor Fluid Handling
Compatible with aggressive chemicals.
- Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
- Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
- Single-use custom lengths and configurations available to minimise downtime
- Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
- Tubing Size: L/S 18
<B>Applications</B>: Chemical, food, beverage, and pharmaceutical applications<br /><B>Characteristics</B>: Excellent chemical resistance, low extractability, low gas permeability, plasticizer free, translucent<br /><br /><B>NOTE:</B> Tygon® Chemical pump tubing is recommended for use in Easy-Load® and Easy-Load® II pump heads only.