Masterflex® Ismatec® Pump Tubing, 2-Stop, Tygon® S3™ E-LFL , 2.79 mm ID, 15" L; 12/PK
Supplier: Avantor Fluid Handling
VWR Item#MFLX06447-48
Your Organization's Price:
Retrieving
Longest life of all Tygon® formulations.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Tubing Inner Diameter: 2.79 mm
For use with Masterflex® L/S® multichannel pump heads.