Masterflex® Ismatec® Pump Tubing, 2-Stop, Tygon® S3™ E-LFL , 0.19 mm ID, 15" L; 12/PK
Supplier: Avantor Fluid Handling
Longest life of all Tygon® formulations.
- Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
- Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
- Single-use custom lengths and configurations available to minimise downtime
- Tubing Inner Diameter: 0.19 mm
For use with Masterflex® L/S® multichannel pump heads.