Calibration weights, OIML R111, class F1
Supplier: ZWIEBEL
E109.44.14A E103.44.14.C E104.44.14.C E105.44.14.C E106.44.14.C E108.44.14.C E109.44.14.C
ZWIEE109.44.14AEA 0 GBP
ZWIEE109.44.14A ZWIEE103.44.14.C ZWIEE104.44.14.C ZWIEE105.44.14.C ZWIEE106.44.14.C ZWIEE108.44.14.C ZWIEE109.44.14.C
Calibration weights, OIML R111, class F1
Weights Calibration Weights
Weights for use in the verification or calibration of class F2 weights and for use with weighing instruments of special accuracy class I and high accuracy class II.
  • Density of 8,0 kg/dm3
  • Magnetic susceptibility of less than 0,01
  • Made of stainless steel
  • Weights with adjustment cavity (except 1 to 20 g monobloc)
Order Now
SPECIFICATIONS
Environmentally Preferable