Reaction vessels, cylindrical, with flat flange
Supplier: witeg Labortechnik
0.992.002
WITG0.992.002EA 937 GBP
WITG0.992.002
Reaction vessels, cylindrical, with flat flange
Reaction Vessels Reaction Vessels, Cylindrical Shape
Borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With flat flange, jacketed, flat bottom.
Order Now
SPECIFICATIONS
Environmentally Preferable