Aluminium seals, lined, WHEATON®
Supplier: DWK Life Sciences
224211-01 224211-06 224211-07 224223-01 224231-01 224235-01 224222-01
218-2102EA 277 GBP
218-2102 WHEA224211-06 WHEA224211-07 218-2109 WHEA224231-01 WHEA224235-01 WHEA224222-01
Aluminium seals, lined, WHEATON®
Closures Chromatography Vial Closures
Open top seal provides for filling or retrieving contents with a syringe, large target diameter.
  • For use with 12×32 mm crimp top vials
  • Choice of liner
They are also ideal for use as closures in chromatography and instrumentation applications.