Blunt needles, Monoject™
Supplier: WESTERN LABORATORY SERVICE
8881202363
WELA8881202363EA 230 GBP
WELA8881202363
Blunt needles, Monoject™
Syringe Needles
The Monoject™ blunt cannula is a sterile, single-use, individually packaged blunt needle.
  • Stainless steel laser welded non-lubricated cannula
  • Aluminium Luer lock hub
  • Easy-grip design makes opening package easy and safe
The tumbled blunt end provides easy entry into pre-pierced reseal port which reduces the possibility of a puncture.