VWR®, Digital Calipers
Supplier: VWR Collection
62379-531
VWRU62379-531EA 219 GBP
VWRU62379-531 819-0012 819-0013
VWR®, Digital Calipers
Calipers
Made from stainless steel. Battery life is about 10000 hours.
  • Ergonomic body and friction thumb roller make readings fast
  • Easy to read, jumbo digit display
  • Moisture and dust resistant steel
Zero button instantly sets closed jaws or any reference point to zero. Floating zero may also be set mid-measurement to read difference results without calculations.
Weight: 255 g.
Delivery information: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by NIST.
Order Now
SPECIFICATIONS
Environmentally Preferable