VWR® VistaVision, Cover Glasses
Supplier: VWR Collection
16004-312
VWRU16004-312EA 0 GBP
VWRU16004-312
VWR® VistaVision, Cover Glasses
Cover Glasses
Microscope cover glasses are made of pure white borosilicate glass with excellent chemical resistance and flatness, and are free of bubbles and scratches.
  • No disturbing self-fluorescence under UV light
  • Suitable for use in coverslippers
Certifications: ISO 8255-1
Delivery information: Rectangular cover glasses No. 1 are supplied 100 per hinged box. Rectangular cover glasses No. 1.5 are supplied 1 oz. per hinged box.