VWR®, Pasteur Pipettes, Disposable
Supplier: VWR Collection
14673-010
VWRU14673-010EA 515 GBP
VWRU14673-010
VWR®, Pasteur Pipettes, Disposable
Pipettes Pasteur/Transfer Pipettes
Borosilicate glass. These disposable pipettes feature fire-polished tops and a straight-cut tip.
  • Smooth, fire-polished tops facilitate bulb attachment
  • Pipettes have an inside diameter of 7 mm, which tapers to a straight-cut tip
  • Positive constriction located 25 mm from the top of the pipette facilitates plugging
Ideal for use in all general laboratory applications, including blood banks, hematology, chemistry, and bacteriology.
Order Now
SPECIFICATIONS
Environmentally Preferable