Copper(II) sulphate pentahydrate 20% w/v aqueous solution

Supplier: VWR Collection
Warning

Synonyms: Cupric sulphate pentahydrate, Copper sulphate pentahydrate

VWRS5998.9020EA 0 GBP
VWRS5998.9020
Copper(II) sulphate pentahydrate 20% w/v aqueous solution
Copper (II) sulphate pentahydrate
Formula: CuSO₄·5H₂O
MW: 249.69 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00149681
CAS Number: 7758-99-8
EINECS: 231-847-6
UN: 3082
ADR: 9 III
Merck Index: 13,02682

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR