Pure-Fit® SIB® Smooth Inner Bore Connector Systems
Supplier: Saint Gobain Life Sciences
PFTC750PP PFR250X500PPAF PF135250PPAF PF135375PPAF PF135500PPAF PFC500PPAF PFLS250PPAF PFLS375PPAF PFLS500PPAF PFMS250PPAF PFMS375PPAF PFMS500PPAF PFT250PPAF PFT375PPAF PFT500PPAF PFX250PPAF PFX375PPAF PFY250PPAF PFY375PPAF PFY500PPAF
VERNPFTC750PPEA 0 GBP
VERNPFTC750PP VERNPFR250X500PPAF VERNPF135250PPAF VERNPF135375PPAF VERNPF135500PPAF VERNPFC500PPAF VERNPFLS250PPAF VERNPFLS375PPAF VERNPFLS500PPAF VERNPFMS250PPAF VERNPFMS375PPAF VERNPFMS500PPAF VERNPFT250PPAF VERNPFT375PPAF VERNPFT500PPAF VERNPFX250PPAF VERNPFX375PPAF VERNPFY250PPAF VERNPFY375PPAF VERNPFY500PPAF
Pure-Fit® SIB® Smooth Inner Bore Connector Systems
Fittings and Connectors
Pure-Fit SIB eliminates entrapment and leak points that can occur with traditional assembly systems. The unique internal design eliminates voids or dead space that can lead to turbulence or stagnation of fluid flow creating the potential for bacterial growth.
  • Total smooth inner bore (SIB) transition design
  • Eliminates entrapment and leak points that occur with traditional fitting systems
  • Complete 360° compression along the entire fitting system
  • No excessive voids at the interface or in the fluid path
  • Utilises the BarbLock® retention and seal system
More Product Information