Tubing, Tygon® 2375
Supplier: Saint Gobain Life Sciences
AJK00002 AJK00004 AJK00003 AJK00007 AJK00009 AJK00012 AJK00017 AJK00022 AJK00027-S AJK00029 AJK00038 AJK00046-S AJK00053 AJK42064
VERNAJK00002EA 202 GBP
VERNAJK00002 VERNAJK00004 VERNAJK00003 VERNAJK00007 VERNAJK00009 VERNAJK00012 VERNAJK00017 VERNAJK00022 VERNAJK00027-S VERNAJK00029 VERNAJK00038 VERNAJK00046-S VERNAJK00053 VERNAJK42064
Tubing, Tygon® 2375
Tubing Transfer Tubing
TYGON® 2375 tubing provides extraordinary chemical resistance in a clear tube.
  • Improved pressure rating by 30 to 50% over its predecessor 2075
  • Non-DEHP for high purity
  • Plasticizer-free for low extractables
  • Clear tubing for easier observation
  • Smoother inner surface
  • Low sorption to minimize cross contamination
  • Industry compliant: FDA, EU Food, Contact 2002/72/EC, NSF 51 food standards, REACH
TYGON® 2375 Ultra chemical resistant tubing truly expands the use of clear tubing where chemical resistance is required. In addition to outstanding chemicalMore Product Information