Tygon® F-4040-A Tubings
Supplier: Saint Gobain Life Sciences
AAG00012 AAG00017 AAG00029 AAG00032 AAG00700 AAG00027 AAG00007 AAG00036 AAG00038 AAG00046 AAG00053 AAG00165
VERNAAG00012EA 161 GBP
VERNAAG00012 VERNAAG00017 VERNAAG00029 VERNAAG00032 VERNAAG00700 VERNAAG00027 VERNAAG00007 VERNAAG00036 VERNAAG00038 VERNAAG00046 VERNAAG00053 VERNAAG00165
Tygon® F-4040-A Tubings
Tubing Transfer Tubing
Tygon® F-4040-A tubing resists the swelling and hardening caused by hydrocarbon-based fluids - significantly reducing the risk of failure due to cracks and leaks. Its minimum extractability safeguards the liquid or vapour being transferred against adulteration. It is extremely flexible and ideal for small engines due to its ability to maneuver tight places and simplify installation.
  • Resists embrittlement
  • Compatible with most hydrocarbons
  • Resistant to swelling
  • Highly flexible, easy to install
  • Specifically developed for fuels and lubricants
  • Ozone and UV
More Product Information