Beverage transfer tubing, Tygon® S3™ B-44-3
Supplier: Saint Gobain Life Sciences
AAB00027 AAB00006 AAB00016 AAB00017 AAB00022 AAB00023 AAB00002 AAB00004 AAB00005 AAB00009 AAB00010 AAB00011 AAB00012 AAB00028 AAB00029 AAB00036 AAB00037 AAB00038
228-1202EA 343 GBP
228-1202 VERNAAB00006 VERNAAB00016 224-0164 VERNAAB00022 VERNAAB00023 VERNAAB00002 VERNAAB00004 VERNAAB00005 VERNAAB00009 VERNAAB00010 VERNAAB00011 224-0163 VERNAAB00028 VERNAAB00029 VERNAAB00036 VERNAAB00037 VERNAAB00038
Beverage transfer tubing, Tygon® S3™ B-44-3
Tubing Transfer Tubing
Formulation B-44-3.
  • Clear as glass for easy visual monitoring of flow
  • Lightweight and flexible for easy, quick installation
  • Broad chemical resistance permits use with virtually all non-solvent chemicals
  • Non-wetting properties allow easy cleaning and complete drainage
  • Contains no BPA or phthalates
Tygon® Beverage tubing is specially formulated for transferring a wide variety of beverages including soft drinks, fruit juices, flavoured teas and bottled water. In virtually all cases, Tygon® Beverage tubing will not affect the taste or odour of productMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable