Label Rolls, Core2Write
Supplier: VELTEK ASSOCIATES
C2WR2X1BLK01 C2WR2X1BLK02 C2WR3X1.5BLK01 C2WR3X1.5BLK02 C2WR3X1BLK01 C2WR3X1BLK02 C2WR4X2BLK01 C2WR4X2BLK02 C2WR4X3BLK01 C2WR4X3BLK02
VELTC2WR2X1BLK01EA 3970 GBP
VELTC2WR2X1BLK01 VELTC2WR2X1BLK02 VELTC2WR3X1.5BLK01 VELTC2WR3X1.5BLK02 VELTC2WR3X1BLK01 VELTC2WR3X1BLK02 VELTC2WR4X2BLK01 VELTC2WR4X2BLK02 VELTC2WR4X3BLK01 VELTC2WR4X3BLK02
Label Rolls, Core2Write
Labels
Constructed using CleanPrint 10, Core2Write labels are extremely durable, smear and smudge proof, chemical resistant, and remain pliable in extreme temperatures. All labels can be customised or left blank.
  • Always retain critical documentation, pre-printed and hand-written
  • Extremely durable
  • Abrasion and tear resistant
  • Smear resistant
  • Can withstand multiple chemicals
  • Can withstand a wide temperature range (–70 to 200 ℃)
  • Fully customisable by size, color, pre-printing
All roll labels come with a three inch plastic core. Can be applied to all types ofMore Product Information