Label Sheets, Core2Write
Supplier: VELTEK ASSOCIATES
C2WR2X1SHEET01 C2WR2X1SHEET02 C2WR3X1SHEET02 C2WR4X2SHEET02 C2WR4X3SHEET02
VELTC2WR2X1SHEET01EA 4820 GBP
VELTC2WR2X1SHEET01 VELTC2WR2X1SHEET02 VELTC2WR3X1SHEET02 VELTC2WR4X2SHEET02 VELTC2WR4X3SHEET02
Label Sheets, Core2Write
Labels
Constructed using CleanPrint 10, Core2Write labels are extremely durable, smear and smudge proof, chemical resistant, and remain pliable in extreme temperatures.
  • Can be applied to sub-zero surfaces
  • Perforation between labels is available
All labels can be customised or left blank.
Ordering information: Also available in sheets (sold separately).
Order Now
SPECIFICATIONS
Environmentally Preferable