Ceramic Magnetic Hotplate Stirrer, AREC 4
Supplier: VELP SCIENTIFIC
F20500670
VELPF20500670EA 420 GBP
VELPF20500670
Ceramic Magnetic Hotplate Stirrer, AREC 4
Stirrers Magnetic Hotplate Stirrers
Robust hotplate stirrer featuring a 10 cm (4") square ceramic top, designed to meet the demands of everyday laboratory applications. The AREC 4 is the entry-level ceramic hotplate stirrer with an aluminium body and technopolymer housing.
  • Brushless motor ensuring power and resistance over time
  • Stirs up to 5 L volume (H₂O) from 100 up to 1500 min⁻¹ (rpm)
  • SpeedServo™ counter reaction technology maintains the speed constant as viscosity changes
  • Advanced heating technology guarantees efficient heating performance
  • The robust 10 cm ceramic top offers exceptional
More Product Information