Gel Documentation Systems, MultiDoc-It™ 130, 3 Door
Supplier: UVP ULTRA VIOLET PRODUCTS
97-0192-05 97-0193-05 97-0194-05 97-0195-05 97-0197-05
UVPA97-0192-05EA 9500 GBP
UVPA97-0192-05 UVPA97-0193-05 UVPA97-0194-05 UVPA97-0195-05 UVPA97-0197-05
Gel Documentation Systems, MultiDoc-It™ 130, 3 Door
Photodocumentation Systems
The MultiDoc-It™ 130 imaging system is designed for multiple user laboratories using fluorescent imaging for capturing gels, plates, and membrane images. The high resolution camera is controlled by UVP image capture and enhancement software.
  • DigiCam 130 digital SLR colour camera with >12 megapixel resolution
  • Sturdy darkroom with interlocked door, light-tight with interior overhead white light, wide access door and UV blocking view port
  • Three door model provides side access doors for cutting or moving gels without opening the front door
  • Capture software
More Product Information