Safety helmets, uvex thermo boss
Supplier: UVEX
9754.300
UVEX9754.300EA 29.8 GBP
UVEX9754.300
Safety helmets, uvex thermo boss
Head Protection Safety Helmets
This uvex thermo boss helmet features a six-point textile band attachment which ensures an optimal fit and high comfort and a conventional suspension harness for comfortable adjustable fit.
  • Made from heat-resistant polycarbonate
  • Side Euroslot adapters
Suitable for use in high ambient temperatures.
Certifications: EN 397, high temperatures (150 °C)