Goggles, Carbonvision
Supplier: UVEX
9307.276 9307.365
UVEX9307.276EA 0 GBP
UVEX9307.276 111-2535
Goggles, Carbonvision
Goggles
Modern materials and manufacturing technologies enable the design of frames which combine reduced size and a high mechanical resistance.
  • TPU layer is permanently fixed to the frame, ensuring a perfect individual fit; easy to clean
  • Lightweight enabling the wearer to work for longer periods without any distractions and problems
  • Flexible soft components adapt perfectly to the face ensuring a perfect fit
  • Convenient clip to remove headband before cleaning
These goggles are recommended for protection against dust, liquid splashes, and mechanical hazards, inMore Product Information