Anti-Transporter Associated Protein Rabbit Polyclonal Antibody

Supplier: US Biological
T8266-05D
USBIT8266-05DEA 533 GBP
USBIT8266-05D
Anti-Transporter Associated Protein Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:10000; Western Blot: 1:1000

Type: Primary
Antigen: Transporter Associated Protein
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR