Anti-MHC Class 1 Chain-related Gene A, B Mouse Monoclonal Antibody (Biotin) [clone: 10F158]

Supplier: US Biological
M3886-75Y2
USBIM3886-75Y2EA 505 GBP
USBIM3886-75Y2
Anti-MHC Class 1 Chain-related Gene A, B Mouse Monoclonal Antibody (Biotin) [clone: 10F158]
Antibodies
Recommended dilution: Flow Cytometry:1:10

Type: Primary
Antigen: MHC Class 1 Chain-related Gene A, B
Clonality: monoclonal
Clone: 10F158
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen name MHC Class 1 Chain-related Gene A, B
Environmentally Preferable
Antibody type Primary
Clonality Monoclonal
Conjugation Biotin
Clone 10F158
Reactivity Human
Host Mouse
Isotype IgG2a
Flow cytometry Yes
Form liquid
Purification purified by Protein G chromatography
Concentration ~0,1 mg/ml
Storage buffer PBS, pH 7,4, 1% BSA, 0,09% sodium azide
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR